accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850567-27-0

IUPAC Name: (3-{[(furan-2-yl)methyl]carbamoyl}phenyl)boronic acid
Synonyms: 3-[(furan-2-ylmethyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 245.04
Molecular Formula: C12H12BNO4
InChi Key: BYRIHRGFMYXTLC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C(=O)NCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results