accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850567-55-4

IUPAC Name: 2-[1-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[1-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 248.1
Molecular Formula: C14H18BFO2
InChi Key: BNROZDXKAQJDJC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C(=C)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results