Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)vinylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH52584MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52584MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52584MD Supplier Thermo Scientific Chemicals Supplier No. H52584MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-55-4
Molecular Formula C14H18BFO2
Molecular Weight (g/mol) 248.10
MDL Number MFCD06659925
InChI Key BNROZDXKAQJDJC-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl vinylboronic acid, pinacol ester, 2-1-4-fluorophenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-4-fluorophenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-fluorophenyl vinylboronic acid pinacol ester, 1-4-fluorophenyl vinylboronic acid,pinacol ester, 4-fluoro-alpha-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene
PubChem CID 44717770
IUPAC Name 2-[1-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C(=C)C1=CC=C(F)C=C1

Specifications

Melting Point 38°C to 42°C
Quantity 250 mg
Formula Weight 248.1
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)vinylboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.