accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850568-08-0

IUPAC Name: (4-ethoxy-3-methylphenyl)boronic acid
Synonyms: 4-ethoxy-3-methylphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: GAFSYSHEDOIUSI-UHFFFAOYSA-N
SMILES: CCOC1=C(C)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results