accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850583-75-4

IUPAC Name: 3,6-bis(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 3,6-bis(thiophen-2-yl)-2H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 300.35
Molecular Formula: C14H8N2O2S2
InChi Key: YIUHGBNJJRTMIE-UHFFFAOYSA-N
SMILES: O=C1NC(C2=CC=CS2)=C2C(=O)NC(C3=CC=CS3)=C12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)