Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Dihydro-3,6-di-2-thienyl-pyrrolo[3,4-c]pyrrole-1,4-dione

Catalog No. 7677435G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-774-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-774-35G Supplier Sigma Aldrich Supplier No. 7677435G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,5-Dihydro-3,6-di-2-thienyl-pyrrolo[3,4-c]pyrrole-1,4-dione is a diketopyrrolopyrrole (DPP) based copolymer and are used widely in thin-film transistors and solar cell devices. This aromatic polymer has electron withdrawing groups and hence is very useful for the synthesis of narrow band gap donor-acceptor polymers which are used as active semiconductors for organic electronics. They give rise to high power conversion efficiency (PCE) in Organic Photovoltaic (OPV) Cells.Used in the synthesis of donor-acceptor polymers which are used in polymer field-effect transistors and bulk heterojunction solar cells.

Specifications

CAS 850583-75-4
Molecular Formula C14H8N2O2S2
Linear Formula C14H8N2O2S2
Quantity 5 g
MDL Number MFCD20257907
Synonym 3,6-Di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2 H,5 H)-dione; DPP
Molecular Weight (g/mol) 300.36
Percent Purity 97%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.