accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850623-40-4

IUPAC Name: potassium trifluoro(4-methanesulfonylphenyl)boranuide
Synonyms: potassium trifluoro(4-methanesulfonylphenyl)boranuide
Molecular Weight (g/mol): 262.09
Molecular Formula: C7H7BF3KO2S
InChi Key: QOZWCMTYKAZMHD-UHFFFAOYSA-N
SMILES: [K+].CS(=O)(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results