accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850661-66-4

IUPAC Name: 3-[(naphthalen-1-yl)methyl]-2,5,5-triphenyl-4-(propan-2-yl)-1,3,2-oxazaborolidine
Synonyms: 4-isopropyl-3-(naphthalen-1-ylmethyl)-2,5,5-triphenyl-1,3,2-oxazaborolidine
Molecular Weight (g/mol): 481.45
Molecular Formula: C34H32BNO
InChi Key: KMDAQDQUTLTEPL-UHFFFAOYNA-N
SMILES: CC(C)C1N(CC2=C3C=CC=CC3=CC=C2)B(OC1(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results