Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-Isopropyl-3-(1-naphthylmethyl)-2,5,5-triphenyl-1,3,2-oxazaborolidine, TCI America™
SDP

Catalog No. I080110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I080110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I080110ML Supplier TCI America Supplier No. I080110ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(ca. 6% in Toluene, ca. 0.1mol/L)

Chemical Identifiers

CAS 850661-66-4
Molecular Formula C34H32BNO
Molecular Weight (g/mol) 481.446
InChI Key KMDAQDQUTLTEPL-XIFFEERXSA-N
PubChem CID 12195798
IUPAC Name (4S)-3-(naphthalen-1-ylmethyl)-2,5,5-triphenyl-4-propan-2-yl-1,3,2-oxazaborolidine
SMILES B1(N(C(C(O1)(C2=CC=CC=C2)C3=CC=CC=C3)C(C)C)CC4=CC=CC5=CC=CC=C54)C6=CC=CC=C6

Specifications

Color Yellow
Quantity 10 mL
UN Number 1294
Formula Weight 481.45
Physical Form Liquid
Chemical Name or Material (S)-4-Isopropyl-3-(1-naphthylmethyl)-2,5,5-triphenyl-1,3,2-oxazaborolidine (ca. 6% in Toluene, ca. 0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.