accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClO

CAS RN 85070-48-0

IUPAC Name: 3-chloro-2-fluorobenzaldehyde
Synonyms: 3-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: YAOZCMANASAVFN-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC=C1C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results