Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluorobenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. C21275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21275G 5 g
C212725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21275G Supplier TCI America Supplier No. C21275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85070-48-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD01631571
InChI Key YAOZCMANASAVFN-UHFFFAOYSA-N
Synonym 2-fluoro-3-chlorobenzaldehyde, 2-fluoro-3-chloro-benzaldehyde, 3-chloro-2-fluoro-benzaldehyde, benzaldehyde, 3-chloro-2-fluoro, pubchem1408, acmc-209q3u, 2-fluoro chloro benzaldehyde, ksc496e9b, 3-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003987
PubChem CID 736335
IUPAC Name 3-chloro-2-fluorobenzaldehyde
SMILES FC1=C(Cl)C=CC=C1C=O

Specifications

Color Yellow
Boiling Point 214°C
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.