CAS RN 85070-67-3
CAS RN 85070-67-3
IUPAC Name:
2-fluoro-4-methylbenzonitrile
Synonyms:
2-fluoro-4-methylbenzonitrile
Molecular Weight (g/mol):
135.14
Molecular Formula:
C8H6FN
InChi Key:
WCGNLBCJPBKXCN-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C#N)C(F)=C1
1
–
1
of
1
results
1
2-Fluoro-4-methylbenzonitrile 98.0+%, TCI America™

CAS: 85070673 Molecular Formula: C8H6FN Molecular Weight (g/mol): 135.14 MDL Number: MFCD03094311 InChI Key: WCGNLBCJPBKXCN-UHFFFAOYSA-N Synonym: 4-cyano-3-fluorotoluene,2-fluoro-4-methylbenzenecarbonitrile,2-fluoro-4-methyl-benzonitrile,benzonitrile, 2-fluoro-4-methyl,pubchem1555,2-fluoro-p-tolunitrile,acmc-209q3w,ksc494e6d,4-methyl-2-fluorobenzonitrile,pharmabridge p-1079 PubChem CID: 2778443 IUPAC Name: 2-fluoro-4-methylbenzonitrile SMILES: CC1=CC=C(C#N)C(F)=C1