Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-methylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. F08845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08845G 5 g
F08841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08845G Supplier TCI America Supplier No. F08845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85070-67-3
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD03094311
InChI Key WCGNLBCJPBKXCN-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorotoluene, 2-fluoro-4-methylbenzenecarbonitrile, 2-fluoro-4-methyl-benzonitrile, benzonitrile, 2-fluoro-4-methyl, pubchem1555, 2-fluoro-p-tolunitrile, acmc-209q3w, ksc494e6d, 4-methyl-2-fluorobenzonitrile, pharmabridge p-1079
PubChem CID 2778443
IUPAC Name 2-fluoro-4-methylbenzonitrile
SMILES CC1=CC=C(C#N)C(F)=C1

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.