accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85073-19-4

IUPAC Name: lithium(1+) phenyl(2,4,6-trimethylbenzoyl)phosphinate
Synonyms: lithium(1+) phenyl(2,4,6-trimethylbenzoyl)phosphinate
Molecular Weight (g/mol): 294.21
Molecular Formula: C16H16LiO3P
InChi Key: JUYQFRXNMVWASF-UHFFFAOYSA-M
SMILES: [Li+].CC1=CC(C)=C(C(=O)P([O-])(=O)C2=CC=CC=C2)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)