accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850852-85-6

IUPAC Name: 1-(1,3-thiazol-2-yl)methanamine hydrochloride
Synonyms: 1-(1,3-thiazol-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 150.62
Molecular Formula: C4H7ClN2S
InChi Key: AAJCCSYSPIKAJY-UHFFFAOYSA-N
SMILES: Cl.NCC1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results