Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-thiazol-2-ylmethylamine, 97%, Thermo Scientific™

Catalog No. CC09513DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC09513DE 5 g Amber Glass Bottle
CC09513DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09513DE Supplier Thermo Scientific Chemicals Supplier No. CC09513DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850852-85-6
Molecular Formula C4H7ClN2S
Molecular Weight (g/mol) 150.624
MDL Number MFCD02854204
InChI Key AAJCCSYSPIKAJY-UHFFFAOYSA-N
Synonym thiazol-2-ylmethanamine hydrochloride, 2-aminomethylthiazole hcl, 1,3-thiazol-2-ylmethanamine hydrochloride, 2-aminomethylthiazole hydrochloride, 2-amino methylthiazole hydrochloride, c-thiazol-2-yl-methylamine hydrochloride, 2-thiazolemethanamine hydrochloride, 1,3-thiazol-2-ylmethylamine hydrochloride, 2-aminomethyl thiazole hydrochloride, 1,3-thiazol-2-ylmethylamine, chloride
PubChem CID 42614216
IUPAC Name 1,3-thiazol-2-ylmethanamine;hydrochloride
SMILES C1=CSC(=N1)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 150.63
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-2-ylmethylamine hydrochloride
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.