accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNOCH3HOOH

CAS RN 850991-69-4

IUPAC Name: (5-methoxypyridin-3-yl)boronic acid
Synonyms: 5-methoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: ISDFOFZTZUILPE-UHFFFAOYSA-N
SMILES: COC1=CN=CC(=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results