CAS RN 85100-78-3
IUPAC Name:
1-hexyl-3-methyl-1H-imidazol-3-ium bromide
Synonyms:
1-hexyl-3-methylimidazolium bromide
Molecular Weight (g/mol):
247.18
Molecular Formula:
C10H19BrN2
InChi Key:
BGSUDDILQRFOKZ-UHFFFAOYSA-M
SMILES:
[Br-].CCCCCCN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1-Hexyl-3-methylimidazolium Bromide 98.0+%, TCI America™
CAS: 85100-78-3 Molecular Formula: C10H19BrN2 Molecular Weight (g/mol): 247.18 MDL Number: MFCD03427612 InChI Key: BGSUDDILQRFOKZ-UHFFFAOYSA-M PubChem CID: 2734237 IUPAC Name: 1-hexyl-3-methylimidazol-3-ium;bromide SMILES: CCCCCCN1C=C[N+](=C1)C.[Br-]