Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyl-3-methylimidazolium Bromide 98.0+%, TCI America™
SDP

Catalog No. H12275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12275G 5 g
H122725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12275G Supplier TCI America Supplier No. H12275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85100-78-3
Molecular Formula C10H19BrN2
Molecular Weight (g/mol) 247.18
MDL Number MFCD03427612
InChI Key BGSUDDILQRFOKZ-UHFFFAOYSA-M
PubChem CID 2734237
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;bromide
SMILES CCCCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Boiling Point 395°C
Quantity 5 g
Formula Weight 247.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.