CAS RN 851232-71-8
CAS RN 851232-71-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with acetone, Pd 13.0-15.0%, Thermo Scientific™
CAS: 851232-71-8 Molecular Formula: C34H28Cl2FeP2Pd·(CH3)2CO MDL Number: MFCD00792899 Synonym: [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride; PdCl2(dppf).(CH3)2CO