accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2NNOOPOOOPOOOPOOOOHOHNaNaNaNa

CAS RN 851265-78-6

IUPAC Name: tetrasodium [({5-[6-amino-2-(2-phenylethynyl)-9H-purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methyl phosphonato)oxy](phosphonatooxy)phosphinate
Synonyms: tetrasodium ({5-[6-amino-2-(2-phenylethynyl)purin-9-yl]-3,4-dihydroxyoxolan-2-yl}methyl phosphonato)oxy(phosphonatooxy)phosphinate
Molecular Weight (g/mol): 695.23
Molecular Formula: C18H16N5Na4O13P3
InChi Key: ONTPGXYSNYMMOK-UHFFFAOYNA-J
SMILES: [Na+].[Na+].[Na+].[Na+].NC1=C2N=CN(C3OC(COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)C(O)C3O)C2=NC(=N1)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results