CAS RN 851335-07-4
IUPAC Name:
4-(dihydroxyboranyl)-3-fluorobenzoic acid
Synonyms:
4-(dihydroxyboranyl)-3-fluorobenzoic acid
Molecular Weight (g/mol):
183.93
Molecular Formula:
C7H6BFO4
InChi Key:
TVNVDRMGOOLVOX-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(F)C=C(C=C1)C(O)=O