Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carboxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C32205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3220-5G 5 g
C32201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3220-5G Supplier TCI America Supplier No. C32205G
Add to Cart
Add to Cart

Specifications

CAS 851335-07-4
Color White
Molecular Formula C7H6BFO4
Quantity 5 g
MDL Number MFCD08458473
Synonym 4-Carboxy-2-fluorobenzeneboronic Acid
InChI Key TVNVDRMGOOLVOX-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C(=O)O)F)(O)O
IUPAC Name 4-borono-3-fluorobenzoic acid
Molecular Weight (g/mol) 183.929
PubChem CID 23134177
Formula Weight 183.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.