accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 851768-62-2

IUPAC Name: 1-imino-1',3'-dihydro-1H-spiro[naphthalene-2,2'-perimidin]-8-amine
Synonyms: 1-imino-1',3'-dihydrospiro[naphthalene-2,2'-perimidin]-8-amine
Molecular Weight (g/mol): 312.38
Molecular Formula: C20H16N4
InChi Key: CXGACCZEFCWAJN-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1C(=N)C1(NC3=CC=CC4=CC=CC(N1)=C34)C=C2
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results