accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3ONOOOOH

CAS RN 85201-91-8

IUPAC Name: 1-[(benzyloxy)carbonyl]-4-(tert-butoxy)pyrrolidine-2-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]-4-(tert-butoxy)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 321.37
Molecular Formula: C17H23NO5
InChi Key: YCTNIMOEFJORHQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC1CC(N(C1)C(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)