accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852180-71-3

IUPAC Name: methyl({[3-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: QSOPQWOPMDNJLF-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=NOC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results