accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONN

CAS RN 852180-74-6

IUPAC Name: 3-(pyrimidin-5-yl)benzoic acid
Synonyms: 3-(pyrimidin-5-yl)benzoic acid
Molecular Weight (g/mol): 200.2
Molecular Formula: C11H8N2O2
InChi Key: URQRFVRGUHUTMI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CN=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results