accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852180-77-9

IUPAC Name: methyl({4-[2-(morpholin-4-yl)ethoxy]phenyl}methyl)amine
Synonyms: methyl({4-[2-(morpholin-4-yl)ethoxy]phenyl}methyl)amine
Molecular Weight (g/mol): 250.34
Molecular Formula: C14H22N2O2
InChi Key: CFDWJPMGCABITD-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OCCN2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results