accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85232-02-6

IUPAC Name: tert-butyl N-(diethoxyphosphoryl)carbamate
Synonyms: tert-butyl N-(diethoxyphosphoryl)carbamate
Molecular Weight (g/mol): 253.24
Molecular Formula: C9H20NO5P
InChi Key: MJJRHRHTDZXUKY-UHFFFAOYSA-N
SMILES: CCOP(=O)(NC(=O)OC(C)(C)C)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results