accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3CNNAuClCH3H3CCH3CH3(?)(?)

CAS RN 852445-83-1

IUPAC Name: {1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene}(chloro)gold
Synonyms: [1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](chloro)gold
Molecular Weight (g/mol): 621.02
Molecular Formula: C27H36AuClN2
InChi Key: CNJQLSINQGKZAW-UHFFFAOYSA-M
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1C=CN(C1=[Au]Cl)C1=C(C=CC=C1C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)