Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) 98.0+%, TCI America™
SDP

Catalog No. C2405200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2405200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2405200MG Supplier TCI America Supplier No. C2405200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 852445-83-1
Molecular Formula C27H36AuClN2
Molecular Weight (g/mol) 621.016
MDL Number MFCD09839143
InChI Key CNJQLSINQGKZAW-UHFFFAOYSA-M
PubChem CID 53338759
IUPAC Name [1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-2-ylidene]-chlorogold
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=CN(C2=[Au]Cl)C3=C(C=CC=C3C(C)C)C(C)C

Specifications

Melting Point 298°C
Color White-Yellow
Quantity 200 mg
Formula Weight 621.02
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.