accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 853073-44-6

IUPAC Name: di-tert-butyl(methyl)phosphanium; tetraphenylboranuide
Synonyms: di-tert-butyl(methyl)phosphanium tetraphenylborate
Molecular Weight (g/mol): 480.48
Molecular Formula: C33H42BP
InChi Key: GPICDLFVQCUBQN-UHFFFAOYSA-O
SMILES: C[PH+](C(C)(C)C)C(C)(C)C.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results