Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butylmethylphosphonium Tetraphenylborate 98.0+%, TCI America™
SDP

Catalog No. D33301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33301G Supplier TCI America Supplier No. D33301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 853073-44-6
Molecular Formula C33H42BP
Molecular Weight (g/mol) 480.48
MDL Number MFCD08276365
InChI Key GPICDLFVQCUBQN-UHFFFAOYSA-O
PubChem CID 23136641
IUPAC Name di-tert-butyl(methyl)phosphanium; tetraphenylboranuide
SMILES C[PH+](C(C)(C)C)C(C)(C)C.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 480.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butylmethylphosphonium Tetraphenylborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.