accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOBr

CAS RN 853234-57-8

IUPAC Name: 2,8-dibromo-6H,12H-indeno[1,2-b]fluorene-6,12-dione
Synonyms: 2,8-dibromoindeno[1,2-b]fluorene-6,12-dione
Molecular Weight (g/mol): 440.09
Molecular Formula: C20H8Br2O2
InChi Key: PFFSYBJLQOJRQU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC4=C(C=C3C(=O)C2=C1)C1=C(C=C(Br)C=C1)C4=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results