Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,8-Dibromoindeno[1,2-b]fluorene-6,12-dione 98.0+%, TCI America™
SDP

Catalog No. D45731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45731G Supplier TCI America Supplier No. D45731G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 853234-57-8
Molecular Formula C20H8Br2O2
Molecular Weight (g/mol) 440.09
MDL Number MFCD22571712
InChI Key PFFSYBJLQOJRQU-UHFFFAOYSA-N
PubChem CID 12173629
IUPAC Name 2,8-dibromoindeno[1,2-b]fluorene-6,12-dione
SMILES C1=CC2=C(C=C1Br)C(=O)C3=CC4=C(C=C23)C(=O)C5=C4C=CC(=C5)Br

Specifications

Quantity 1 g
Formula Weight 440.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dibromoindeno[1,2-b]fluorene-6,12-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.