CAS RN 85326-10-9
IUPAC Name:
N-[9-(5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl]-2-methylpropanamide
Synonyms:
N-[9-(5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl)-6-oxo-3H-purin-2-yl]-2-methylpropanamide
Molecular Weight (g/mol):
451.6
Molecular Formula:
C20H33N5O5Si
InChi Key:
WZQWRHWLQPEBFF-UHFFFAOYNA-N
SMILES:
CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(CO[Si](C)(C)C(C)(C)C)O1
1
–
1
of
1
results
1
Promotions Available
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
CAS: 85326-10-9 Molecular Formula: C20H33N5O5Si Molecular Weight (g/mol): 451.599 MDL Number: MFCD04972287 InChI Key: WZQWRHWLQPEBFF-BFHYXJOUSA-N Synonym: n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine,n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide PubChem CID: 12016034 IUPAC Name: N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide SMILES: CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O