Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%

Catalog No. AAH5272606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5272606 5 g
AAH5272603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5272606 Supplier Thermo Scientific Chemicals Supplier No. H5272606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 85326-10-9 | C20H33N5O5Si | 451.599 g/mol

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85326-10-9
Molecular Formula C20H33N5O5Si
Molecular Weight (g/mol) 451.599
MDL Number MFCD04972287
InChI Key WZQWRHWLQPEBFF-BFHYXJOUSA-N
Synonym n2-isobutyryl-5'-o-tert-butyldimethylsilyl-2'-deoxyguanosine, n-9-2r,4s,5r-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide
PubChem CID 12016034
IUPAC Name N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO[Si](C)(C)C(C)(C)C)O

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 451.6
Percent Purity 97%
Chemical Name or Material N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.