CAS RN 85328-80-9
IUPAC Name:
4-aminonaphthalene-1,5-disulfonic acid sodium
Synonyms:
4-aminonaphthalene-1,5-disulfonic acid sodium
Molecular Weight (g/mol):
326.29
Molecular Formula:
C10H9NNaO6S2
InChi Key:
ABPYYYVZWBYEAE-UHFFFAOYSA-N
SMILES:
[Na].NC1=CC=C(C2=CC=CC(=C12)S(O)(=O)=O)S(O)(=O)=O