Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 4-Amino-1,5-naphthalenedisulfonate 98.0+%, TCI America™
SDP

Catalog No. A034225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A034225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A034225G Supplier TCI America Supplier No. A034225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85328-80-9
Molecular Formula C10H9NNaO6S2
Molecular Weight (g/mol) 326.293
MDL Number MFCD00021519
InChI Key ABPYYYVZWBYEAE-UHFFFAOYSA-N
Synonym 1-Naphthylamine-4,8-disulfonic Acid Monosodium Salt, 4-Amino-1,5-naphthalenedisulfonic Acid Monosodium Salt, Monosodium 4-Amino-1,5-naphthalenedisulfonate
PubChem CID 86734804
IUPAC Name 4-aminonaphthalene-1,5-disulfonic acid;sodium
SMILES C1=CC2=C(C=CC(=C2C(=C1)S(=O)(=O)O)N)S(=O)(=O)O.[Na]

Specifications

Quantity 25 g
Formula Weight 325.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Amino-1,5-naphthalenedisulfonate
EINECSNumber (2)-0502
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.