accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3POCH3OCH3OCH3OH3C

CAS RN 85417-41-0

IUPAC Name: tris(2,6-dimethoxyphenyl)phosphane
Synonyms: tris(2,6-dimethoxyphenyl)phosphane
Molecular Weight (g/mol): 442.45
Molecular Formula: C24H27O6P
InChi Key: CMLWFCUAXGSMBB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1P(C1=C(OC)C=CC=C1OC)C1=C(OC)C=CC=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results