accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CCH3NNPOCH3H3CCH3CH3

CAS RN 854929-36-5

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-oxo-1,3,2λ⁵-diazaphospholidin-2-ylium
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-2-oxo-1,3,2λ⁵-diazaphospholidin-2-ylium
Molecular Weight (g/mol): 425.58
Molecular Formula: C26H38N2OP
InChi Key: PJGJPWAXZUCDHL-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1CCN(C2=C(C=CC=C2C(C)C)C(C)C)[P+]1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results