Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,6-diisopropylphenyl)-1,3,2-diazaphospholidine 2-Oxide 98.0+%, TCI America™
SDP

Catalog No. B34791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34791G Supplier TCI America Supplier No. B34791G
Add to Cart
Add to Cart

Specifications

CAS 854929-36-5
Melting Point 203°C
Color White-Yellow
Molecular Formula C26H38N2OP+
Quantity 1 g
InChI Key PJGJPWAXZUCDHL-UHFFFAOYSA-N
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2CCN([P+]2=O)C3=C(C=CC=C3C(C)C)C(C)C
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-1,3,2-diazaphospholidin-2-ium 2-oxide
Molecular Weight (g/mol) 425.577
PubChem CID 12183214
Formula Weight 426.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)-1,3,2-diazaphospholidine 2-Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.