CAS RN 855153-75-2
CAS RN 855153-75-2
IUPAC Name:
2-(piperidin-1-yl)pyridine-4-carboxylic acid
Synonyms:
2-(piperidin-1-yl)pyridine-4-carboxylic acid
Molecular Weight (g/mol):
206.25
Molecular Formula:
C11H14N2O2
InChi Key:
PYHYIVXTQSCHDX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=NC=C1)N1CCCCC1
1
–
1
of
1
results
1
2-Piperidinoisonicotinic acid, 97%, Thermo Scientific™
CAS: 855153-75-2 Molecular Formula: C11H14N2O2 Molecular Weight (g/mol): 206.245 MDL Number: MFCD04115214 InChI Key: PYHYIVXTQSCHDX-UHFFFAOYSA-N Synonym: 2-piperidinoisonicotinic acid,2-piperidin-1-yl isonicotinic acid,2-piperidin-1-yl pyridine-4-carboxylic acid,2-piperidin-1-ylisonicotinic acid,2-piperidin-1-ylpyridin-4-ylcarboxylic acid,3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4'-carboxylic acid,4-pyridinecarboxylic acid, 2-1-piperidinyl,2-piperidylpyridine-4-carboxylic acid,2-piperidin-1-yl-isonicotinic acid,1-4-carboxypyridin-2-yl piperidine PubChem CID: 6484237 IUPAC Name: 2-piperidin-1-ylpyridine-4-carboxylic acid SMILES: C1CCN(CC1)C2=NC=CC(=C2)C(=O)O