Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperidinoisonicotinic acid, 97%, Thermo Scientific™

Catalog No. CC50301DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC50301DA 1 g Amber Glass Bottle
CC50301CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC50301DA Supplier Thermo Scientific Chemicals Supplier No. CC50301DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 855153-75-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD04115214
InChI Key PYHYIVXTQSCHDX-UHFFFAOYSA-N
Synonym 2-piperidinoisonicotinic acid, 2-piperidin-1-yl isonicotinic acid, 2-piperidin-1-yl pyridine-4-carboxylic acid, 2-piperidin-1-ylisonicotinic acid, 2-piperidin-1-ylpyridin-4-ylcarboxylic acid, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4'-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-piperidinyl, 2-piperidylpyridine-4-carboxylic acid, 2-piperidin-1-yl-isonicotinic acid, 1-4-carboxypyridin-2-yl piperidine
PubChem CID 6484237
IUPAC Name 2-piperidin-1-ylpyridine-4-carboxylic acid
SMILES C1CCN(CC1)C2=NC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 2-Piperidinoisonicotinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.