accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOFFFOCH3FFF

CAS RN 85541-57-7

IUPAC Name: 3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl 3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
Synonyms: 3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl 3,3,3-trifluoro-2-methoxy-2-phenylpropanoate
Molecular Weight (g/mol): 450.33
Molecular Formula: C20H16F6O5
InChi Key: AATFPUCRBIXXID-UHFFFAOYNA-N
SMILES: COC(C(=O)OC(=O)C(OC)(C1=CC=CC=C1)C(F)(F)F)(C1=CC=CC=C1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results