CAS RN 856169-60-3
CAS RN 856169-60-3
IUPAC Name:
2-chloro-3,5-diiodopyridine
Synonyms:
2-chloro-3,5-diiodopyridine
Molecular Weight (g/mol):
365.34
Molecular Formula:
C5H2ClI2N
InChi Key:
FVCKBKBSDFLXGY-UHFFFAOYSA-N
SMILES:
ClC1=C(I)C=C(I)C=N1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-Chloro-3,5-diiodopyridine, 95%, Thermo Scientific™
CAS: 856169-60-3 Molecular Formula: C5H2ClI2N Molecular Weight (g/mol): 365.34 MDL Number: MFCD04966987 InChI Key: FVCKBKBSDFLXGY-UHFFFAOYSA-N Synonym: pyridine,2-chloro-3,5-diiodo PubChem CID: 632570 IUPAC Name: 2-chloro-3,5-diiodopyridine SMILES: ClC1=C(I)C=C(I)C=N1