accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 856412-22-1

OOOHONH(S)H2C

CAS RN 856412-22-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hept-6-enoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hept-6-enoic acid
Molecular Weight (g/mol): 365.43
Molecular Formula: C22H23NO4
InChi Key: LRSIYGFZZWFAAI-FQEVSTJZSA-N
SMILES: OC(=O)[C@H](CCCC=C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)