accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85650-56-2

CH3N(R)(R)ClOOHOOHO(Z)

CAS RN 85650-56-2

IUPAC Name: (2R,6R)-9-chloro-4-methyl-13-oxa-4-azatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),7,9,11,14,16-hexaene; (2Z)-but-2-enedioic acid
Synonyms: asenapine; maleic acid
Molecular Weight (g/mol): 401.84
Molecular Formula: C21H20ClNO5
InChi Key: GMDCDXMAFMEDAG-TXUHOWFZSA-N
SMILES: OC(=O)\C=C/C(O)=O.CN1C[C@@H]2[C@@H](C1)C1=CC(Cl)=CC=C1OC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results