accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOOOOOOOHNaNaNa

CAS RN 85715-60-2

IUPAC Name: trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms: edta trisodium|edetate trisodium|edta trisodium salt|trisodium ethylenediaminetetraacetate|ethylenediaminetetraacetic acid trisodium salt|trisodium salt of ethylenediaminetetraacetic acid|(ethylenedinitrilo)tetraacetic acid trisodium salt
Molecular Weight (g/mol): 358.19
Molecular Formula: C10H13N2Na3O8
InChi Key: QZKRHPLGUJDVAR-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Physical Form 
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (1)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (7)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)