Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%

Catalog No. AC147865000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC147865000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147865000 Supplier Thermo Scientific Chemicals Supplier No. 147865000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 85715-60-2 | C10H13N2Na3O8 | 358.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 85715-60-2
Melting Point 300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (corrected for water) (Complexometry)
Packaging Plastic bottle
Molecular Formula C10H13N2Na3O8
Linear Formula (NaO2CCH2)2NCH2CH2N(CH2CO2Na)CH2CO2H·xH2O
Quantity 500 g
MDL Number MFCD00149675
Synonym trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
Solubility Information Solubility in water: soluble.
InChI Key QZKRHPLGUJDVAR-UHFFFAOYSA-K
SMILES [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;hydrate
Molecular Weight (g/mol) 358.19
PubChem CID 16211179
Formula Weight 394.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.